Få mere at vide
Saxagliptin hydrochloride, Tocris Bioscience™
GREENER_CHOICE
DPPIV/CD26 Inhibitors
Brand:  Tocris Bioscience 6507/10
Encompass
Produktkode 15997960
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Saxagliptin hydrochloride is a high affinity DPP-IV inhibitor (Ki = 0.6 nM). Inhibits plasma DPP-IV (87%). Elicits enhanced glucose clearance in Zuckerfa/fa rats.
Tekniske data
Tekniske data
709031-78-7
(1S,3S,5S)-2-[(2S)-Amino(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile hydrochloride
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner