Få mere at vide
Saxagliptin Hydrochloride, LoGiCal
Artikelnummer. 30461414
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30461414 10 mg
1 options
Artikelnummer. 30461414 Leverandør LoGiCal Leverandørnr. LGCFOR3377.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Saxagliptin Hydrochloride
CAS 709031-78-7
Analyt- eller komponentnavne Saxagliptin Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C18 H25 N3 O2 . Cl H
Certificeringer/compliance ISO 9001
Synonym 2-Azabicyclo[3.1.0]hexane-3-carbonitrile, 2-[(2S)-2-amino-2-(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-, hydrochloride (1:1), (1S,3S,5S)-, 2-Azabicyclo[3.1.0]hexane-3-carbonitrile, 2-[(2S)-amino(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-, monohydrochloride, (1S,3S,5S)- (9CI), Saxagliptin hydrochloride
InChI formel InChI=1S/C18H25N3O2.ClH/c19-8-13-2-12-3-14(12)21(13)16(22)15(20)17-4-10-1-11(5-17)7-18(23,6-10)9-17;/h10-15,23H,1-7,9,20H2;1H/t10?,11?,12-,13+,14+,15-,17?,18?;/m1./s1
Løsningstype Neat
SMIL Cl.N[C@H](C(=O)N1[C@@H](C[C@@H]2C[C@H]12)C#N)C34CC5CC(CC(O)(C5)C3)C4
IUPAC navn (1S,3S,5S)-2-[(2S)-2-amino-2-(3-hydroxy-1-adamantyl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 351.87
Mængde 10 mg
Formel vægt 351.1714 g/mol