Få mere at vide
Savolitinib, TRC
Artikelnummer. 30416105
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30416105 5 mg
30412746 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30416105 Leverandør Toronto Research Chemicals Leverandørnr. TRCS1429035MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Savolitinib
CAS 1313725-88-0
Molekylær formel C17H15N9
Mængde 5 mg
Synonym Volitinib, 1-[(1S)-1-Imidazo[1,2-a]pyridin-6-ylethyl]-6-(1-methyl-1H-pyrazol-4-yl)-1H-1,2,3-triazolo[4,5-b]pyrazine, AZD 6094, HM 5016504, HMPL 504
InChI formel InChI=1S/C17H15N9/c1-11(12-3-4-15-18-5-6-25(15)10-12)26-17-16(22-23-26)19-8-14(21-17)13-7-20-24(2)9-13/h3-11H,1-2H3/t11-/m0/s1
SMIL C[C@@H](c1ccc2nccn2c1)n3nnc4ncc(nc34)c5cnn(C)c5
IUPAC navn 3-[(1S)-1-imidazo[1,2-a]pyridin-6-ylethyl]-5-(1-methylpyrazol-4-yl)triazolo[4,5-b]pyrazine
Molekylvægt (g/mol) 345.36
Formel vægt 345.145