Learn More
Thermo Scientific™ Savant™ SpeedVac™ Rotors for Medium Capacity Concentrators
Product Code. 11758589 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
10 Centrifuge Tubes
100 (6 x 50mm) Tubes
100 (8 x 29 mm) Tubes
12 Scintillation Vials
12 Vials
24 Vials
32 Tubes
4 x 100 mL Pear-shaped Flasks
40 Microtubes
40 Tubes
6 x 50 mL Conical Tubes
60 Vials
64 Microtubes
72 Tubes
8 (17.5 x 102 mm, 15mL Corex) Tubes
8 (18 x 100, 17 x 95, 16 x 100 mm) Tubes
Unit Size:
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
11758589 8 (17.5 x 102 mm, 15mL Corex) Tubes Each
11758609 100 (6 x 50mm) Tubes Each
11949426 100 (8 x 29 mm) Tubes Each
11738629 10 Centrifuge Tubes Each
11798539 12 Vials Each
11768609 12 Scintillation Vials Each
11796427 24 Vials Each
11798589 24 Vials Each
11768659 32 Tubes Each
11748539 40 Microtubes Each
11738539 40 Tubes Each
11738619 4 x 100 mL Pear-shaped Flasks Each
11708549 60 Vials Each
11718549 64 Microtubes Each
11728629 6 x 50 mL Conical Tubes Each
11788629 72 Tubes Each
10433332 8 (18 x 100, 17 x 95, 16 x 100 mm) Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 11758589 Supplier Thermo Scientific™ Supplier No. RH817.5
  • ONLINE EXCLUSIVE PRICE
    9250.00 DKK
    8654.71 DKK / Pakke med 1
    Spare 595.29 DKK
    6% Slukket
View all promotions
 Restricted Item
Forventet forsendelse: 16-03-2027
Select from one of these rotors to fit the type of sample volume and various sample formats for your specific application. Choose from various size of tubes, flasks, mircrowell plates.
SpeedVac Rotors for Medium Capacity Concentrators for use with Savant SpeedVac Concentrators.
Circular Rotors:
  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
TRUSTED_SUSTAINABILITY
Specifications
Description Glass and Plastic Tubes: 8 (17.5 x 102mm, 15mL Corex), Medium capacity models
Material Glass (Tube), Plastic (Tube)
Type Rotor
For Use With (Equipment) SPD1030, SPD120, SPD130, SPD140DDA
Holds 8 (17.5 x 102 mm, 15mL Corex) Tubes
Model RH8-17.5
Rotor System Traditional
Volume (Metric) 15 mL