Få mere at vide
Satraplatin, MedChemExpress
Artikelnummer. 16487376
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16487376 100 mg -
16459877 5 mg -
16417408 10 mg 10mg
16403437 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16487376 Leverandør Medchem Express Leverandørnr. HY17576
MedChemExpress Satraplatin is an alkylating agent, with potent antitumor effect.
Satraplatin is an alkylating agent, with potent antitumor effect.
Tekniske data
Kemisk navn eller materiale Satraplatin
Fysisk form Solid
CAS 129580-63-8
Farve Off-White
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C10H22Cl2N2O4Pt
Til brug med (applikation) Neuroscience-Neuromodulation
Synonym BMS182751 BMY45594 JM216
Opløselighedsinformation DMSO : ≥ 33 mg/mL (65.96 mM; DMSO can inactivate Satraplatin's activity) ∣DMF : 10 mg/mL (19.99 mM; ultrasonic and adjust pH to 1 with HCl; DMSO can inactivate Satraplatin's activity) ∣Ethanol : < 1 mg/mL (insoluble; DMSO can inactivate Satraplatin's activity) ∣H2O : < 0.1 mg/mL (insoluble; DMSO can inactivate Satraplatin's activity)
SMIL [Cl-][Pt+4]([O-]C(C)=O)([O-]C(C)=O)([Cl-])([NH3])[NH2]C1CCCCC1
Molekylvægt (g/mol) 500.28
Formel vægt 500.28
Procent renhed 98.05%