missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Quantitative Grade 389 Filter Papers

Brand:  Sartorius FT-3-102-110

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10438362

  • 182.00 DKK / Pack of 100


Estimated Shipment
29-04-2021
Add to basket

Description

Description

  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications

Specifications

White
Disc
8 to 12μm
0.19 mm
110 mm
389
100/Pk.
Videos
SDS
Documents