Få mere at vide
Sartorius Entris™ Toploading Balances
Shop alle Sartorius produkter
Volumen:
820 g
2.2 kg
3.2 kg
4.2 kg
5.2 kg
6.2 kg
8.2 kg
Læsbarhed:
Inkluderer:
External Calibration
Internal Calibration
Pakningsstørrelse:
Hver
Beskrivelse
- White-backlit, high-contrast display with 15 mm digits; exceptionally easy to read under any room lighting conditions
- New operating concept: easy configuration of the individual settings guided by short, menu-driven text prompts (choice of 5 languages) and simple navigation using cursor keys
- RS-232C bidirectional interface port; optional Sartorius cable for connection of the balance to a USB port
- Total ease of operation
- All keys have positive click action
- Clearly defined control panel
- Monolithic weighing cell provides high thermal stability and extremely fast response times
- Fast response time and display stability
- Language-driven user interface
- Unit toggle of up to 5 units
Applications
Percent weighing, unit toggle, animal weighing, net-total, calculation, density determination and counting
Innocal Calibration Services
Available in items containing i-1SUS in Manufacturer's No.: 14-559-023, -024,- 025, -026, -027, -028, -029, -030.
Tekniske data
Tekniske data
| Tilslutningstype | RS-232C, 25-pin |
| Volumen | 5.2 kg |
| Læsbarhed | 100 mg |
| Inkluderer | Internal Calibration |
| Linearitet | 300 mg |
| Model- | ENTRIS5201i-1S |
| Gentagelighed | 100 mg |
| Stabiliseringstid | 1.5 sec. |
| Type | Toploading Balance |
| Dimensioner (L x B x H) | 303 x 230 x 91 mm (12.9 x 9.0 x 3.6 in.) |
| Vis mere |