Learn More
Sartorius Quantitative Grade 388 Filter Papers
These filter papers are used for quantitative and gravimetric analyses as well as for pressure or vacuum filtration. They are made out of 100 % cotton linters with an α-cellulose content of > 98 % and are acid-washed to make the papers ashless and achieve high purity.
Brand:  Sartorius FT-3-101-400
Additional Details : Weight : 1.33500kg
 View more versions of this product
Product Code. 15982840
  • 2505.00 DKK / Pack of 100
Estimated Shipment: 09-07-2024
to see stock.
Add to basket
Description
Description
Quantitative Grade 388 Filter Papers
  • Ash-free (ash content ≤ 0.01 % according to DIN 540370)
  • Wet-strengthened
  • Color-coded box for easy selection
Specifications
Specifications
Black
10 to 15 μm
388
0.21 mm
Disc
10 sec.
100/Pk.
400 mm