Få mere at vide
Sars Envelope Protein Rabbit anti-SARS-CoV, Polyclonal, Novus Biologicals™
Product Code. 18193739 Shop All Bio Techne Products
Change view
Click to view available options
Mængde:
0.1 mg
Product Code. Mængde
18193739 0.1 mg
1 options
Product Code. 18193739 Supplier Novus Biologicals Supplier No. NBP2410620.1MG
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Rabbit Polyclonal Antibody
Sars Envelope Protein Polyclonal antibody specifically detects Sars Envelope Protein in SARS-CoV samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Antigen Sars Envelope Protein
Applikationer ELISA
Klassifikation Polyclonal
Koncentration 1 mg/mL
Konjugeret Unconjugated
Fortynding ELISA 1 ug/mL
Formulering PBS
Gene Alias SARS coronavirus Envelope protein, SARS CoV E protein, SARS CoV Envelope protein, SARSCoV, SARSCoV Envelope protein, Severe acute respiratory syndrome
Værtsarter Rabbit
Immunogen Anti-Sars Envelope Protein antibody was raised against a peptide corresponding to 11 amino acids near the carboxy terminus of SARS-CoV Envelope protein. The immunogen is located within the last 50 amino acids of SARS Envelope Proteins.
Oprensningsmetode Peptide affinity purified
Mængde 0.1 mg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 1489671
Målarter SARS-CoV
Indhold og opbevaring Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG