missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 Spike Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12851AF594
Additional Details : Weight : 0.00970kg
Description
SARS-CoV-2 Spike Polyclonal antibody specifically detects SARS-CoV-2 Spike in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
SARS-CoV-2 Spike | |
Polyclonal | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
Affinity-purified | |
RUO | |
Primary | |
SARS-CoV-2 | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
Alexa Fluor 594 | |
2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein | |
Anti-SARS-CoV-2 Spike L452R (Delta Variant) antibody was raised against a peptide corresponding to 9 amino acids in the receptor binding domain of SARS-CoV-2 Delta variant (B.1.617.2) Spike protein. The immunogen is located within 430-480 amino acids of SARS-CoV-2 Delta variant (B.1.617.2) Spike protein. | |
0.1 mL | |
Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
43740568 | |
Store at 4°C in the dark. | |
IgG |