missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ SARS-CoV-2 Nucleocapsid Recombinant Rabbit Monoclonal Antibody (84D7), Invitrogen™
GREENER_CHOICE

Artikelnummer. 17156798 Shop alle Thermo Scientific produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17156798 200 μL
17137048 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17156798 Leverandør Invitrogen™ Leverandørnr. MA537209

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Recombinant Monoclonal Antibody

SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen SARS-CoV-2 Nucleocapsid
Applikationer ELISA
Klassifikation Recombinant Monoclonal
Klon 84D7
Koncentration 0.5 mg/mL
Konjugeret Unconjugated
Formulering PBS with 0.02% sodium azide
Gene N
Genadgangsnr. P0DTC9
Gene Alias COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N
Gensymboler N
Værtsarter Rabbit
Immunogen SARS-CoV-2 nucleoprotein (NP) specific peptide.
Oprensningsmetode Protein A
Mængde 200 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 43740575
Målarter Virus
Indhold og opbevaring 4°C
Produkttype Antibody
Form Liquid
Isotype IgG κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.