missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SARS-CoV-2 nsp2 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-07051FR
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SARS-CoV-2 nsp2 Polyclonal antibody specifically detects SARS-CoV-2 nsp2 in SARS-CoV-2 samples. It is validated for Western Blot, ELISA, Immunofluorescence
Tekniske data
| SARS-CoV-2 nsp2 | |
| Polyclonal | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Rabbit | |
| Peptide Affinity-purified | |
| RUO | |
| Primary | |
| SARS-CoV-2 | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 680 | |
| Non-structural protein 2, NSP2, p65 homolog | |
| This antibody was raised against a peptide corresponding to 14 amino acids near the center of SARS-CoV-2 (COVID-19) NSP2 protein. The immunogen is located within 90-130 amino acids of SARS-CoV-2 (COVID-19) NSP2 protein. | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 43740578 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion