missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SARS-CoV-2 nsp2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-07052AF405
Additional Details : Weight : 0.00970kg
Description
SARS-CoV-2 nsp2 Polyclonal antibody specifically detects SARS-CoV-2 nsp2 in SARS-CoV-2 samples. It is validated for ELISA, Immunohistochemistry, ImmunofluorescenceSpecifications
SARS-CoV-2 nsp2 | |
Polyclonal | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Rabbit | |
Peptide Affinity-purified | |
RUO | |
Primary | |
SARS-CoV-2 | |
Purified |
ELISA, Immunohistochemistry, Immunofluorescence | |
Alexa Fluor 405 | |
Non-structural protein 2, NSP2, p65 homolog | |
This antibody was raised against a peptide corresponding to 15 amino acids near the center of SARS-CoV-2 (COVID-19) NSP2 protein. The immunogen is located within 50-100 amino acids of the SARS-CoV-2 (COVID-19) NSP2 protein. | |
0.1 mL | |
Infections (Virus Bacteria and Parasites) | |
43740578 | |
Store at 4°C in the dark. | |
IgG |