Learn More
SARS-CoV-2 Membrane Recombinant Rabbit Monoclonal Antibody (HL1087), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA546923
Additional Details : Weight : 0.01000kg
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The coronavirus membrane protein (M) is the most abundant viral structural protein and plays a central role in virus assembly and morphogenesis|M protein forms a mushroom-shaped dimer, composed of two transmembrane domain-swapped three-helix bundles and two intravirion domains|M protein further assembles into higher-order oligomers|A highly conserved hinge region is key for conformational changes|The M protein dimer is unexpectedly similar to SARS-CoV-2 ORF3a, a viral ion channel|The interaction analyses of M protein with nucleocapsid protein (N) and RNA suggest that the M protein mediates the concerted recruitment of these components through the positively charged intravirion domain [PMID: 35931673]Specifications
SARS-CoV-2 Membrane | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
HL1087 | |
Unconjugated | |
CoV-2; SARS | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of SARS-CoV-2 (COVID-19) Membrane. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |