missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ SARS Coronavirus Matrix Synthetic Peptide
Klik for at se tilgængelige muligheder
Mængde:
50 μg
Pakningsstørrelse:
50µg
Beskrivelse
SARS Matrix Synthetic Peptide, PEP-0260, from Invitrogen.
A novel coronavirus has recently been identified as the causative agent of SARS (Severe Acute Respiratory Syndrome). Coronaviruses are a major cause of upper respiratory diseases in humans. The genomes of these viruses are positive-stranded RNA approximately 27-31kb in length. The M protein (Membrane protein, Matrix protein) is one of the major structural viral proteins. It is an integral membrane protein involved in the budding of the viral particles and interacts with S (Spike) protein and the nucleocapsid protein.
Tekniske data
Tekniske data
| Arter | Virus |
| Oprensningsmetode | purified |
| Fælles navn | SARS Coronavirus Matrix |
| Koncentration | 200 μg/mL |
| Indhold og opbevaring | -20°C |
| Oversigt | Synthetic Peptide |
| Format | Liquid |
| Formulering | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| Til brug med (applikation) | Blocking Assay, Control |
| Lyofiliseret, frysetørret | No |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion