Få mere at vide
Sarpogrelate Hydrochloride, TRC
Artikelnummer. 30423494
Skift visning
Mængde:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423494 100 mg
30413521 250 mg
30418059 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30423494 Leverandør Toronto Research Chemicals Leverandørnr. TRCS142300100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 135159-51-2
Molekylær formel C24H31NO6.HCl
Molekylvægt (g/mol) 465.97
Synonym 1-[2-(Dimethylamino)-1-[[2-[2-(3-methoxyphenyl)ethyl]phenoxy]methyl]ethyl]butanedioic Acid Ester Hydrochloride (1:1), Butanedioic acid, mono[2-(dimethylamino)-1-[[2-[2-(3-methoxyphenyl)ethyl]phenoxy]methyl]ethyl] ester, hydrochloride (9CI), Butanedioic acid, mono[2-(dimethylamino)-1-[[2-[2-(3-methoxyphenyl)ethyl]phenoxy]methyl]ethyl] ester, hydrochloride, (±)-, Anplag, MCI 9042, Sarpogrelate hydrochloride
IUPAC navn 4-[1-(dimethylamino)-3-[2-[2-(3-methoxyphenyl)ethyl]phenoxy]propan-2-yl]oxy-4-oxobutanoic acid;hydrochloride
SMIL Cl.COc1cccc(CCc2ccccc2OCC(CN(C)C)OC(=O)CCC(=O)O)c1
Tekniske data
Kemisk navn eller materiale Sarpogrelate Hydrochloride
Mængde 100 mg
InChI formel InChI=1S/C24H31NO6.ClH/c1-25(2)16-21(31-24(28)14-13-23(26)27)17-30-22-10-5-4-8-19(22)12-11-18-7-6-9-20(15-18)29-3;/h4-10,15,21H,11-14,16-17H2,1-3H3,(H,26,27);1H
Formel vægt 465.192