Få mere at vide
Sarcosine-d3, TRC
Artikelnummer. 30180085
Skift visning
Mængde:
25 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180085 25 mg
30179920 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30180085 Leverandør Toronto Research Chemicals Leverandørnr. TRCS14050425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 118685-91-9
Molekylær formel C3H4D3NO2
Molekylvægt (g/mol) 92.11
Synonym N-(Methyl-d3)-glycine, N-(Methyl-d3)glycine, [2H3]Sarcosine
SMIL OC(CNC([2H])([2H])[2H])=O
Tekniske data
Kemisk navn eller materiale Sarcosine-d3
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C3H7NO2/c1-4-2-3(5)6/h4H,2H2,1H3,(H,5,6)/i1D3
Formel vægt 92.0665
Procent renhed >95%