Få mere at vide
Sarcosine-d3 Hydrochloride, TRC
Artikelnummer. 30425810
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30425810 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425810 Leverandør Toronto Research Chemicals Leverandørnr. TRCS14050225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sarcosine-d3 Hydrochloride
CAS 347840-04-4
Molekylær formel C3 2H3 H4 N O2 . Cl H
Mængde 25 mg
Synonym Glycine','N-(methyl-d3)-','hydrochloride (9CI), N-(Methyl-d3)glycine hydrochloride, Sarcosine-methyl-d3 hydrochloride, [2H3]Sarcosine hydrochloride, N-Methylglycine-D3 (N-methyl-D3) hydrochloride, Sarcosine-d3 (N-methyl-d3) hydrochloride
InChI formel InChI=1S/C3H7NO2.ClH/c1-4-2-3(5)6;/h4H,2H2,1H3,(H,5,6);1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])NCC(=O)O
IUPAC navn 2-(trideuteriomethylamino)acetic acid;hydrochloride
Molekylvægt (g/mol) 128.57
Formel vægt 128.0432