Få mere at vide
Sarafloxacin Hydrochloride, LoGiCal
Artikelnummer. 30459985
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30459985 10 mg
1 options
Artikelnummer. 30459985 Leverandør LoGiCal Leverandørnr. LGCFOR3452.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sarafloxacin hydrochloride
CAS 91296-87-6
Analyt- eller komponentnavne Sarafloxacin hydrochloride
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C20 H17 F2 N3 O3 . Cl H
Certificeringer/compliance ISO 9001
Synonym 6-Fluoro-1-(4-fluorophenyl)-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid hydrochloride (1:1), 3-Quinolinecarboxylic acid, 6-fluoro-1-(4-fluorophenyl)-1,4-dihydro-4-oxo-7-(1-piperazinyl)-, monohydrochloride (9CI), Sarafin, Sarafloxacin hydrochloride
InChI formel InChI=1S/C20H17F2N3O3.ClH/c21-12-1-3-13(4-2-12)25-11-15(20(27)28)19(26)14-9-16(22)18(10-17(14)25)24-7-5-23-6-8-24;/h1-4,9-11,23H,5-8H2,(H,27,28);1H
Løsningstype Neat
SMIL Cl.OC(=O)C1=CN(c2ccc(F)cc2)c3cc(N4CCNCC4)c(F)cc3C1=O
IUPAC navn 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 421.82
Mængde 10 mg
Formel vægt 421.1005 g/mol