Få mere at vide
SAR 7334, Tocris Bioscience™
Potent TRPC6 inhibitor
Brand: Tocris Bioscience 5831/5
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
SAR 7334 is a potent TRPC6 inhibitor (IC50 values are 9.5, 226 and 282 nM for TRPC6, TRPC7 and TRPC3-mediated Ca2+ influx). Exhibits no significant activity at TRPC4 and TRPC5 channels. Suppresses TRPC6-dependent acute hypoxic pulmonary vasoconstriction (HPV) in the isolated perfused mouse lung. Orally bioavailable.Tekniske data
| 4-[[(1R,2R)-2-[(3R)-3-Amino-1-piperidinyl]-2,3-dihydro-1H-inden-1-yl]oxy]-3-chlorobenzonitrile dihydrochloride | |
| 1333207-63-8 | |
| >98% | |
| 5 mg |