Få mere at vide
Tocris Bioscience™ Saquinavir mesylate
GREENER_CHOICE
Artikelnummer. 18075322
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18075322 10 mg
18095832 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18075322 Leverandør Tocris Bioscience™ Leverandørnr. 4418/10
HIV protease inhibitor
Saquinavir mesylate is an inhibitor of human immunodeficiency virus (HIV) protease (Ki values are <0.1 and 0.12 nM for HIV-2 and HIV-1 protease respectively). Exhibits high antiviral activity and low cytotoxicity. In silico docking models predict potential as an inhibitor of SARS-CoV-2 3CLpro.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 149845-06-7
Molekylær formel C39H54N6O8S
Tekniske data
Kemisk navn eller materiale Saquinavir mesylate
Mængde 10 mg
Formel vægt 766.95
Renhed >99%
Molekylvægt (g/mol) 766.955