Få mere at vide
Skytec™ Sapphire Total Gloves
Artikelnummer. p-200110044
Størrelse:
Small
Medium
Large
X-Large
2X-Large
Artikelnummer. 30274529
Brand: Skytec™ SKY00121
Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
A great fitting, fully coated glove combining high cut protection and excellent grip in oily and wet conditions. Ideal for oily tasks with sharp parts that require great grip.
  • Oil Resistant Water Repellent Cut Resistant Grip Glove
  • Cut resistant: Level E/ ANSI A5
  • A flexible
  • No irritation by seams in contact with the skin
  • Elasticated wrist provides a secure fit
Tekniske data
Certificeringer/overholdelse (Europa) CE II, ANSI/ISEA 105-2016: Cut A5, EN 388:2016 + A1:2018 4X43E, EN 407:2020 X1XXXX, EN 21420:2020
Konstruktion Seamless Knit
Belægningsmateriale Foam Nitrile
Farve Black
Bedømmelse af skæremodstand Level E
Hånd Ambidextrous
Latex fri Yes
Materiale Steel Fiber
Produkttype Cut-resistant Gloves
Størrelse Small
EN388 Standard 4X43E
Liner materiale HPPE, Nylon, Spandex
Længde (metrisk) 230 mm