Få mere at vide
Santalol, TRC
Artikelnummer. 17912165
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17912165 100 mg
17941295 50 mg
17911725 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17912165 Leverandør Toronto Research Chemicals Leverandørnr. TRCS115018100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 11031-45-1
Molekylær formel C15 H24 O
Molekylvægt (g/mol) 220.351
IUPAC navn (E)-2-methyl-5-(2-methyl-3-methylidene-2-bicyclo[2.2.1]heptanyl)pent-2-en-1-ol
SMIL C\C(=C/CCC1(C)C2CCC(C2)C1=C)\CO
Tekniske data
Kemisk navn eller materiale Santalol
Mængde 100 mg
InChI formel InChI=1S/C15H24O/c1-11(10-16)5-4-8-15(3)12(2)13-6-7-14(15)9-13/h5,13-14,16H,2,4,6-10H2,1,3H3/b11-5+
Formel vægt 220.183