missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
SAMSN1 Antibody [mFluor Violet 500 SE], Novus Biologicals Biologicals™
Shop alle Bio Techne produkterBeskrivelse
SAMSN1 Polyclonal antibody specifically detects SAMSN1 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Tekniske data
Tekniske data
| Antigen | SAMSN1 |
| Applikationer | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Klassifikation | Polyclonal |
| Konjugeret | mFluor Violet 500 SE |
| Formulering | 50mM Sodium Borate |
| Gene Alias | HACS1gene with homology to KIAA0790 protein10NASH1, Hematopoietic adaptor containing SH3 and SAM domains 1, SAM and SH3 domain containing 1, SAM and SH3 domain containing 2, SAM domain, SH3 domain and nuclear localisation signals, 1, SAM domain, SH3 domain and nuclear localization signals 1, SAM domain, SH3 domain and nuclear localization signals protein 1, SAM domain-containing protein SAMSN-1, SH3D6B, SH3-SAM adaptor protein |
| Værtsarter | Rabbit |
| Immunogen | SAMSN1 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human SAMSN. The immunogen is located within the last 50 amino acids of SAMSN. |
| Oprensningsmetode | Peptide affinity purified |
| Mængde | 0.1 mL |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?