Få mere at vide
Salmeterol D3, Dr. Ehrenstorfer
Artikelnummer. 30319796
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30319796 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319796 Leverandør Dr. Ehrenstorfer Leverandørnr. DREC16904910
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Salmeterol-d3
CAS 497063-94-2
Molekylær formel C25 2H3 H34 N O4
Mængde 10 mg
InChI formel InChI=1S/C25H37NO4/c27-20-23-18-22(13-14-24(23)28)25(29)19-26-15-7-1-2-8-16-30-17-9-6-12-21-10-4-3-5-11-21/h3-5,10-11,13-14,18,25-29H,1-2,6-9,12,15-17,19-20H2/i20D2,25D
SMIL [2H]C([2H])(O)c1cc(ccc1O)C([2H])(O)CNCCCCCCOCCCCc2ccccc2
IUPAC navn 4-[1-deuterio-1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]-2-[dideuterio(hydroxy)methyl]phenol
Molekylvægt (g/mol) 418.58
Formel vægt 418.2911 g/mol