Få mere at vide
(±)-Salmeterol-d3 (3-hydroxymethyl-d2; alpha-d1), CDN
Artikelnummer. 30320216
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320216 0.005 g
30309904 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320216 Leverandør CDN Leverandørnr. CDND61870.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Salmeterol-d3
CAS 497063-94-2
Molekylær formel C25 2H3 H34 N O4
Mængde 0.005 g
InChI formel InChI=1 S/C25H37NO4/c27-20-23-18-22(13-14-24(23)28)25(29)19-26-15-7-1-2-8-16-30-17-9-6-12-21-10-4-3-5-11-21/h3-5,10-11,13-14,18,25-29 H,1-2,6-9,12,15-17,19-20H2/i20D2,25 D
SMIL [2 H]C([2 H])(O)c1cc(ccc1O)C([2 H])(O)CNCCCCCCOCCCCc2ccccc2
IUPAC navn 4-[1-deuterio-1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]-2-[dideuterio(hydroxy)methyl]phenol
Molekylvægt (g/mol) 418.58
Formel vægt 418.2911 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity