Få mere at vide
Salinazid, LoGiCal
Artikelnummer. 30464183
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464183 10 mg
1 options
Artikelnummer. 30464183 Leverandør LoGiCal Leverandørnr. LGCFOR1962.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Salinazid
CAS 495-84-1
Analyt- eller komponentnavne Salinazid
Forsendelsestilstand Room Temperature
Molekylær formel C13 H11 N3 O2
Certificeringer/compliance ISO 9001
Synonym 4-Pyridinecarboxylic acid, 2-[(2-hydroxyphenyl)methylene]hydrazide, 4-Pyridinecarboxylic acid, [(2-hydroxyphenyl)methylene]hydrazide (9CI), Isonicotinic acid, salicylidenehydrazide (6CI,7CI,8CI), Hydrazine, 1-isonicotinoyl-2-salicylidene- (6CI), (2-Hydroxybenzylidene)pyridine-4-carbohydrazide, 1-Isonicotinoyl-2-salicylidenehydrazine, INSH, N-Isonicotinamidosalicylaldimine, N-Isonicotinoyl-N'-(salicylidene)hydrazine, NSC 33760, Neobicina Salicilica, Nilazid, Nupasal, Nupasal 213, Phthisosan, SAH, SIH, Salicylaldehyde 4-pyridinecarboxylic acid hydrazone, Salicylaldehyde isonicotinoylhydrazine, Salicylaldehyde isonicotinoylhydrazone, Salicylidene isonicotinoylhydrazone, Salinazid, Salizid, Salizide, o-Hydroxybenzal isonicotinylhydrazone, o-Hydroxybenzaldehyde isonicotinoylhydrazone, o-Oxyberon
InChI formel InChI=1S/C13H11N3O2/c17-12-4-2-1-3-11(12)9-15-16-13(18)10-5-7-14-8-6-10/h1-9,17H,(H,16,18)
Løsningstype Neat
SMIL Oc1ccccc1C=NNC(=O)c2ccncc2
IUPAC navn N-[(2-hydroxyphenyl)methylideneamino]pyridine-4-carboxamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 241.25
Mængde 10 mg
Formel vægt 241.0851 g/mol