missing translation for 'onlineSavingsMsg'
Få mere at vide

Salicyloyl Phytosphingosine, TRC

Artikelnummer. 30412379
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412379 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30412379 Leverandør Toronto Research Chemicals Leverandørnr. TRCS63448010MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Salicyloyl Phytosphingosine
CAS 212908-67-3
Molekylær formel C25 H43 N O5
Mængde 10 mg
Synonym Salicyloyl Phytosphingosine, N-[(1S,2S,3R)-2,3-Dihydroxy-1-(hydroxymethyl)heptadecyl]-2-hydroxybenzamide
InChI formel InChI=1S/C25H43NO5/c1-2-3-4-5-6-7-8-9-10-11-12-13-18-23(29)24(30)21(19-27)26-25(31)20-16-14-15-17-22(20)28/h14-17,21,23-24,27-30H,2-13,18-19H2,1H3,(H,26,31)/t21-,23+,24-/m0/s1
SMIL CCCCCCCCCCCCCC[C@@H](O)[C@@H](O)[C@H](CO)NC(=O)c1ccccc1O
IUPAC navn 2-hydroxy-N-[(2S,3S,4R)-1,3,4-trihydroxyoctadecan-2-yl]benzamide
Molekylvægt (g/mol) 437.613
Formel vægt 437.314
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.