Få mere at vide
Salicylic Acid Acyl Glucoside, TRC
Artikelnummer. 30135093
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135093 25 mg
30133112 2.5 mg
30005789 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135093 Leverandør Toronto Research Chemicals Leverandørnr. TRCS08811525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 60517-74-0
Molekylær formel C13H16O8
Molekylvægt (g/mol) 300.26
Synonym Glucosyl Salicylate, 1-(2-Hydroxybenzoate) β-D-Glucopyranose;
SMIL OC1=C(C(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)=O)C=CC=C1
Tekniske data
Kemisk navn eller materiale Salicylic Acid Acyl Glucoside
Mængde 25 mg
InChI formel InChI=1S/C13H16O8/c14-5-8-9(16)10(17)11(18)13(20-8)21-12(19)6-3-1-2-4-7(6)15/h1-4,8-11,13-18H,5H2/t8-,9-,10+,11-,13+/m1/s1
Formel vægt 300.0845