Få mere at vide
Salicylic Acid Acyl-Beta-D-glucuronide, TRC
Artikelnummer. 30136844
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136844 100 mg
30006269 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136844 Leverandør Toronto Research Chemicals Leverandørnr. TRCS088135100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 29315-53-5
Molekylær formel C13 H14 O9
Molekylvægt (g/mol) 314.24
Synonym beta-D-Glucopyranuronic acid, 1-(2-hydroxybenzoate), Glucopyranosiduronic acid, 1-salicylate, beta-D- (8CI), Salicyl acyl glucuronide, Salicylic acid glucuronide, Salicylic acid, beta-D-glucopyranuronosyl ester
IUPAC navn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(2-hydroxybenzoyl)oxyoxane-2-carboxylic acid
SMIL O[C@@H]1[C@@H](O)[C@H](OC(=O)c2ccccc2O)O[C@@H]([C@H]1O)C(=O)O
Tekniske data
Kemisk navn eller materiale Salicylic Acid Acyl-beta-D-glucuronide
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C13H14O9/c14-6-4-2-1-3-5(6)12(20)22-13-9(17)7(15)8(16)10(21-13)11(18)19/h1-4,7-10,13-17H,(H,18,19)/t7-,8-,9+,10-,13-/m0/s1
Formel vægt 314.0638
Procent renhed >95%