missing translation for 'onlineSavingsMsg'
Få mere at vide

Salicylic Acid 2-O-beta-D-Glucoside-D4, TRC

Artikelnummer. 30408844
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30408844 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408844 Leverandør Toronto Research Chemicals Leverandørnr. TRCS08816425MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Salicylic Acid 2-O-beta-D-Glucoside-D4
CAS 1174218-98-4
Molekylær formel C13 D4 H12 O8
Mængde 25 mg
Synonym 6-(β-D-Glucopyranosyloxy)benzoic-2,3,4,5-D4 Acid, 2-(((2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzoic-3,4,5,6-D4 Acid, o-(Glucopyranosyloxy)benzoic Acid-D4, o-(β-D-Glucopyranosyloxy)benzoic Acid-D4, o-(β-D-Glucosyloxy)benzoic Acid-D4, 2-(β-D-Glucopyranosyloxy)benzoic Acid-D4, 2-O-β-D-Glucosylsalicylic Acid-D4, 2-O-β-Glucopyranosylsalicylic Acid-D4, Salicylate 2-O-β-D-Glucoside-D4, Salicylic Acid 2-β-D-Glucoside-D4, Salicylic Acid O-Glucoside-D4
InChI formel InChI=1S/C13H16O8/c14-5-8-9(15)10(16)11(17)13(21-8)20-7-4-2-1-3-6(7)12(18)19/h1-4,8-11,13-17H,5H2,(H,18,19)/t8-,9-,10+,11-,13-/m1/s1/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c(C(=O)O)c(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)c1[2H]
IUPAC navn 2,3,4,5-tetradeuterio-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzoic acid
Molekylvægt (g/mol) 304.286
Formel vægt 304.11
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.