missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Salicylic Acid 2-O-beta-D-Glucoside-D4, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Salicylic Acid 2-O-beta-D-Glucoside-D4 |
| CAS | 1174218-98-4 |
| Molekylær formel | C13 D4 H12 O8 |
| Mængde | 2.5 mg |
| Synonym | 6-(β-D-Glucopyranosyloxy)benzoic-2,3,4,5-D4 Acid, 2-(((2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)benzoic-3,4,5,6-D4 Acid, o-(Glucopyranosyloxy)benzoic Acid-D4, o-(β-D-Glucopyranosyloxy)benzoic Acid-D4, o-(β-D-Glucosyloxy)benzoic Acid-D4, 2-(β-D-Glucopyranosyloxy)benzoic Acid-D4, 2-O-β-D-Glucosylsalicylic Acid-D4, 2-O-β-Glucopyranosylsalicylic Acid-D4, Salicylate 2-O-β-D-Glucoside-D4, Salicylic Acid 2-β-D-Glucoside-D4, Salicylic Acid O-Glucoside-D4 |
| InChI formel | InChI=1S/C13H16O8/c14-5-8-9(15)10(16)11(17)13(21-8)20-7-4-2-1-3-6(7)12(18)19/h1-4,8-11,13-17H,5H2,(H,18,19)/t8-,9-,10+,11-,13-/m1/s1/i1D,2D,3D,4D |
| SMIL | [2H]c1c([2H])c([2H])c(C(=O)O)c(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)c1[2H] |
| IUPAC navn | 2,3,4,5-tetradeuterio-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxybenzoic acid |
| Molekylvægt (g/mol) | 304.286 |
| Formel vægt | 304.11 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?