missing translation for 'onlineSavingsMsg'
Få mere at vide

Salcaprozate Sodium, TRC

Artikelnummer. 30418800
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418800 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30418800 Leverandør Toronto Research Chemicals Leverandørnr. TRCS085600500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Salcaprozate Sodium
CAS 203787-91-1
Renhedsgrad noter HPLC
Molekylær formel C15 H20 N O4 . Na
Mængde 500 mg
Synonym Octanoic acid','8-[(2-hydroxybenzoyl)amino]-','sodium salt (1:1), Octanoic acid','8-[(2-hydroxybenzoyl)amino]-','monosodium salt (9CI), E 414, SNAC, Salcaprozate sodium, Sodium 8-(salicyloylamino)octanoate, Sodium 8-[(2-hydroxybenzoyl)amino]caprylate, Sodium 8-[(2-hydroxybenzoyl)amino]octanoate, Sodium N-[8-(2-hydroxybenzoyl)amino]caprylate, 8-[(2-Hydroxybenzoyl)amino]octanoic acid sodium salt, 8-[(2-Hydroxybenzoyl)amino]octanoic Acid Monosodium Salt
InChI formel InChI=1S/C15H21NO4.Na/c17-13-9-6-5-8-12(13)15(20)16-11-7-3-1-2-4-10-14(18)19;/h5-6,8-9,17H,1-4,7,10-11H2,(H,16,20)(H,18,19);/q;+1/p-1
SMIL [Na+].Oc1ccccc1C(=O)NCCCCCCCC(=O)[O-]
IUPAC navn sodium;8-[(2-hydroxybenzoyl)amino]octanoate
Molekylvægt (g/mol) 301.31
Formel vægt 301.129
Procent renhed >95
Vis mere Vis mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.