Få mere at vide
Salbutamon-d9 Hydrochloride, TRC
Artikelnummer. 30420033
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30420033 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420033 Leverandør Toronto Research Chemicals Leverandørnr. TRCS0855021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Salbutamon-d9 Hydrochloride
CAS 1346605-08-0
Molekylær formel C13 D9 H10 N O3 . Cl H
Mængde 1 mg
Synonym 2-[(1,1-Dimethylethyl-d9)amino]-1-[4-hydroxy-3-(hydroxymethyl)phenyl]ethanone Hydrochloride
InChI formel InChI=1S/C13H19NO3.ClH/c1-13(2,3)14-7-12(17)9-4-5-11(16)10(6-9)8-15;/h4-6,14-16H,7-8H2,1-3H3;1H/i1D3,2D3,3D3;
SMIL Cl.[2H]C([2H])([2H])C(NCC(=O)c1ccc(O)c(CO)c1)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1-[4-hydroxy-3-(hydroxymethyl)phenyl]-2-[[2,2,2-trideuterio-1,1-bis(trideuteriomethyl)ethyl]amino]ethanone;hydrochloride
Molekylvægt (g/mol) 282.81
Formel vægt 282.1697