Få mere at vide
Salbutamol Acetonide, TRC
Artikelnummer. 30415902
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30415902 50 mg
30421737 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30415902 Leverandør Toronto Research Chemicals Leverandørnr. TRCS08553050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 54208-72-9
Molekylær formel C16 H25 N O3
Molekylvægt (g/mol) 279.37
Synonym α-[[(1,1-Dimethylethyl)amino]methyl]-2,2-dimethyl-4H-1,3-benzodioxin-6-methanol, 2-(tert-Butylamino)-1-(2,2-dimethyl-1,3-benzodioxin-6-yl)ethanol, Salbutamol Acetonide
IUPAC navn 2-(tert-butylamino)-1-(2,2-dimethyl-4H-1,3-benzodioxin-6-yl)ethanol
SMIL CC(C)(C)NCC(O)c1ccc2OC(C)(C)OCc2c1
Tekniske data
Kemisk navn eller materiale Salbutamol Acetonide
Mængde 50 mg
InChI formel InChI=1S/C16H25NO3/c1-15(2,3)17-9-13(18)11-6-7-14-12(8-11)10-19-16(4,5)20-14/h6-8,13,17-18H,9-10H2,1-5H3
Formel vægt 279.1834