Få mere at vide
Saint-Gobain Chemware™ PTFE Watch Glasses
Diameter (metrisk):
5 cm
Kapacitet (metrisk):
11 ml
Pakningsstørrelse:
Hver
Beskrivelse
Used for weighing, crystallization, and drying
Chemically compatible with virtually all industrial chemicals and solvents; exceptions include molten alkali metals, fluorine, and strong fluorinating agents.
- All sizes molded to same contour for easy stacking
- Noncontaminating, nonstick
- Also act as beaker covers for Chemware PTFE Griffin Beakers (05-586-1 Series)
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 11 ml |
| Materiale | PTFE |
| Form | Round |
| Type | Watch Glass |
| Til brug med (applikation) | Crystallization and Drying, Used as Beaker Covers, Weighing |
| Til brug med (udstyr) | For 50mL beaker |
| Diameter (engelsk) | 1.96 in. |
| Diameter (metrisk) | 5 cm |