Få mere at vide
Saikosaponin A, TRC
Artikelnummer. 30417253
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417253 5 mg
30427120 25 mg
30410220 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30417253 Leverandør Toronto Research Chemicals Leverandørnr. TRCS0839005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 20736-09-8
Molekylær formel C42 H68 O13
Molekylvægt (g/mol) 780.98
Synonym β-D-Galactopyranoside, (3β,4α,16β)-13,28-epoxy-16,23-dihydroxyolean-11-en-3-yl 6-deoxy-3-O-β-D-glucopyranosyl-, Saikosaponin A (8CI), (3β,4α,16β)-13,28-Epoxy-16,23-dihydroxyolean-11-en-3-yl 6-deoxy-3-O-β-D-glucopyranosyl-β-D-galactopyranoside, Oleanane, β-D-galactopyranoside deriv., Salikosaponin A
SMIL C[C@H]1O[C@@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@H]3CCC4(C)C(CCC5(C)C4C=CC67OCC8(CCC(C)(C)CC68)[C@@H](O)CC57C)[C@]3(C)CO)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Tekniske data
Kemisk navn eller materiale Saikosaponin A
Mængde 5 mg
InChI formel InChI=1S/C42H68O13/c1-21-28(46)30(48)31(49)34(52-21)55-33-29(47)22(18-43)53-35(32(33)50)54-27-10-11-37(4)23(38(27,5)19-44)8-12-39(6)24(37)9-13-42-25-16-36(2,3)14-15-41(25,20-51-42)26(45)17-40(39,42)7/h9,13,21-35,43-50H,8,10-12,14-20H2,1-7H3/t21-,22-,23?,24?,25?,26+,27+,28-,29+,30+,31-,32-,33+,34+,35+,37?,38+,39?,40?,41?,42?/m1/s1
Formel vægt 780.466