Få mere at vide
Safranal (>80%), TRC
Artikelnummer. 30008374
Skift visning
Mængde:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008374 10 g
30012182 25 g
30003932 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008374 Leverandør Toronto Research Chemicals Leverandørnr. TRCS08190010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 116-26-7
Molekylær formel C10 H14 O
Molekylvægt (g/mol) 150.22
Synonym 2,6,6-Trimethyl-1,3-cyclohexadiene-1-carboxaldehyde, 2,2,6-Trimethyl-4,6-cyclohexadien-1-aldehyde, 2,3-Dihydro-2,2,6-trimethylbenzaldehyde, 2,6,6-Trimethyl-1,3-cyclohexadiene-1-aldehyde, 2,6,6-Trimethylcyclohexa-1,3-diene-1-carbaldehyde, 2,6,6-Trimethylcyclohexa-1,3-dienecarbaldehyde, 2,6,6-Trimethylcyclohexa-1,3-dienecarboxaldehyde, Safranal, 1,3-Cyclohexadiene-1-carboxaldehyde, 2,6,6-trimethyl-
IUPAC navn 2,6,6-trimethylcyclohexa-1,3-diene-1-carbaldehyde
SMIL CC1=C(C=O)C(C)(C)CC=C1
Tekniske data
Kemisk navn eller materiale Safranal
Renhedsgrad noter HPLC
Mængde 10 g
InChI formel InChI=1S/C10H14O/c1-8-5-4-6-10(2,3)9(8)7-11/h4-5,7H,6H2,1-3H3
Formel vægt 150.1045
Procent renhed >95%