Få mere at vide
Safinamide Mesylate, TRC
Artikelnummer. 30133502
Skift visning
Mængde:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133502 100 mg
30135760 50 mg
30008009 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133502 Leverandør Toronto Research Chemicals Leverandørnr. TRCS819080100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 202825-46-5
Molekylær formel C17 H19 F N2 O2 . C H4 O3 S
Molekylvægt (g/mol) 398.45
Synonym Propanamide, 2-[[[4-[(3-fluorophenyl)methoxy]phenyl]methyl]amino]-, (2S)-, methanesulfonate (1:1), Propanamide, 2-[[[4-[(3-fluorophenyl)methoxy]phenyl]methyl]amino]-, (2S)-, monomethanesulfonate (9CI), Propanamide, 2-[[[4-[(3-fluorophenyl)methoxy]phenyl]methyl]amino]-, (S)-, monomethanesulfonate, (S)-2-[[4-[(3-Fluorobenzyl)oxy]benzyl]amino]propanamide methanesulfonate, EMD 1195686 mesylate, NW 1015, PNU 151774E, Safinamide mesilate, Safinamide mesylate, Safinamide methanesulfonate
IUPAC navn (2S)-2-[[4-[(3-fluorophenyl)methoxy]phenyl]methylamino]propanamide;methanesulfonic acid
SMIL C[C@H](NCc1ccc(OCc2cccc(F)c2)cc1)C(=O)N.CS(=O)(=O)O
Tekniske data
Kemisk navn eller materiale Safinamide Mesylate
Mængde 100 mg
InChI formel InChI=1S/C17H19FN2O2.CH4O3S/c1-12(17(19)21)20-10-13-5-7-16(8-6-13)22-11-14-3-2-4-15(18)9-14;1-5(2,3)4/h2-9,12,20H,10-11H2,1H3,(H2,19,21);1H3,(H,2,3,4)/t12-;/m0./s1
Formel vægt 398.1312