Learn More
DWK Life Sciences Safety Stopcock, NS 24/29, for safety outlets type Wertex
The safety stopcock is used for airtight sealing of a desiccator.
Brand: DWK Life Sciences 247960304
434.15 DKK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.06900kg
Description
The safety stopcock is used for airtight sealing of a desiccator. The stopcock can be firmly connected to the desiccator by means of the matching washer blade.
- Tight closure due to standard ground joint
- The safety stopcock can be permanently mounted by means of a sheath
- For safety outlets of WERTEX type desiccator
Specifications
Borosilicate Glass 3.3, Polypropylene | |
Closure of desiccators | |
76 mm |