Learn More
Brady™ Safety Sign COVID-19 Reception
An efficient solution with clear safety instructions to easily communicate COVID-19 countermeasures on work floors.
36.50 DKK - 148.00 DKK
Specifications
Material | Laminated Polyester |
---|---|
Mounting Type | Self-adhesive |
Dimensions (L x W) | 262 x 371 mm (W x H) |
Product Code | Brand | Languages | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Languages | Price | Quantity & Availability | |||||
16506404
|
Brady™
306809 |
Spanish |
146.00 DKK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16516404
|
Brady™
306810 |
Italian |
146.00 DKK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16516444
|
Brady™
306850 |
Norwegian |
36.50 DKK
Each |
In Stock
Log in to see stock. |
|||||
16566394
|
Brady™
306805 |
Dutch |
144.00 DKK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16566424
|
Brady™
306835 |
Danish |
146.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
16576394
|
Brady™
306806 |
French |
148.00 DKK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16576424
|
Brady™
306836 |
Finnish |
146.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
16586394
|
Brady™
306807 |
German |
148.00 DKK
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16586424
|
Brady™
306837 |
Swedish |
146.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
16596394
|
Brady™
306808 |
English |
148.00 DKK
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
Description
- Provides clear, visual communication where it matters
- Highlights the use of appropriate personal protection
- Easily communicates social distancing and hygiene
Specifications
Laminated Polyester | |
Self-adhesive | |
262 x 371 mm (W x H) |