Få mere at vide
Corrosive Resistant Acid and Alkali Cabinets
Dimensioner (D x B x H):
300 x 610 x 610 mm
459 x 459 x 610 mm
459 x 459 x 915 mm
459 x 915 x 712 mm
459 x 915 x 915 mm
459 x 915 x 1220 mm
459 x 915 x 1525 mm
459 x 915 x 1830 mm
Pakningsstørrelse:
Hver
Beskrivelse
- Constructed of 0.9mm-thick zintec steel
- Adjustable spillage and liquid-tight sump tray
- Doors are rebated and secured by chrome three-way locking handle
- White, high-visibility powder-coated finish with appropriate warning labels
- Note: These cabinets are for storing acids safely and do not claim to be acid-proof.
- Cabinets are compliant with the following:
- DSEAR 2002
- HS(G)51, 1990
- Factory Inspectorate Certificate of Approval No1: Parts 3 and 4
- COSHH
Tekniske data
Tekniske data
| Dimensioner (D x B x H) | 459 x 459 x 610 mm |
| Materiale | Steel |
| Produkttype | Acids and Alkalis Corrosive-resistant Cabinet |