Få mere at vide
Sacubitril-d4, TRC
Artikelnummer. 30412188
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30412188 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412188 Leverandør Toronto Research Chemicals Leverandørnr. TRCS08089710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Sacubitril succinate-d4
CAS 1884269-07-1
Renhedsgrad noter HPLC
Molekylær formel C24 2H4 H25 N O5
Mængde 10 mg
Synonym [1,1'-Biphenyl]-4-pentanoic acid','γ-[(3-carboxy-1-oxopropyl-2,2,3,3-d4)amino]-α-methyl-','α-ethyl ester','(αR,γS)-, (2R,4S)-4-[(3-Carboxy-1-oxopropyl-2,2,3,3-d4)amino]-4-[(p-phenylphenyl)methyl]-2-methylbutanoic acid ethyl ester, (2R,4S)-5-(Biphenyl-4-yl)-4-[(3-carboxypropionyl-2,2,3,3-d4)amino]-2-methylpentanoic acid ethyl ester, AHU 377-d4, Sacubitril-d4, Sacubitril-D4 (succinate-D4), Sucabitril-d4
InChI formel InChI=1S/C24H29NO5/c1-3-30-24(29)17(2)15-21(25-22(26)13-14-23(27)28)16-18-9-11-20(12-10-18)19-7-5-4-6-8-19/h4-12,17,21H,3,13-16H2,1-2H3,(H,25,26)(H,27,28)/t17-,21+/m1/s1/i13D2,14D2
SMIL [2H]C([2H])(C(=O)O)C([2H])([2H])C(=O)N[C@@H](C[C@@H](C)C(=O)OCC)Cc1ccc(cc1)c2ccccc2
IUPAC navn 2,2,3,3-tetradeuterio-4-[[(2S,4R)-5-ethoxy-4-methyl-5-oxo-1-(4-phenylphenyl)pentan-2-yl]amino]-4-oxobutanoic acid
Molekylvægt (g/mol) 415.52
Formel vægt 415.2297
Procent renhed >95