Få mere at vide
Saccharin Sodium, Mikromol™
Beskrivelse
Saccharin Sodium
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Saccharin sodium |
| CAS | 128-44-9 |
| Analyt- eller komponentnavne | Saccharin sodium |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C7 H4 N O3 S . Na |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Saccharin Sodium, 1,2-Benzisothiazol-3(2H)-one 1,1-dioxide sodium, 2-Sodio-1,2-benzisothiazol-3(2H)-one 1,1-dioxide, 1,2-Benzisothiazol-3(2H)-one, 1,1-dioxide, sodium salt (1:1), 1,2-Benzisothiazol-3(2H)-one, 1,1-dioxide, sodium salt (9CI), 1,2-Benzisothiazolin-3-one, 1,1-dioxide, sodium deriv. (7CI), 1,2-Benzisothiazolin-3-one, 1,1-dioxide, sodium salt (8CI), Saccharin, sodium deriv. (6CI), 1,2-Benzothiazol-3(2H)-one 1,1-dioxide sodium salt, Cristallose, Crystallose, Kristallose, Saccharin sodium salt, Saccharin soluble, Saxin, Sodium o-benzosulfimide, Sodium o-sulfobenzimide, Sodium saccharide, Sodium saccharin, Sodium saccharinate, Sodium saccharine, Soluble saccharin, Sucram C 150, Sweeta, Sykose, Willosetten, o-Benzoylsulfimide sodium salt |
| InChI formel | InChI=1S/C7H5NO3S.Na/c9-7-5-3-1-2-4-6(5)12(10,11)8-7;/h1-4H,(H,8,9);/q;+1/p-1 |
| SMIL | [Na+].O=C1[N-]S(=O)(=O)c2ccccc12 |
| IUPAC navn | sodium;1,1-dioxo-1,2-benzothiazol-2-id-3-one |
| Vis mere |