Få mere at vide
Saccharin Calcium Hydrate (4:2:7), TRC
Artikelnummer. 30132588
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132588 10 g
30010029 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132588 Leverandør Toronto Research Chemicals Leverandørnr. TRCS08080510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 6381-91-5
Molekylær formel 4 C7 H4 N O3 S . 2 Ca . 7 H2 O
Molekylvægt (g/mol) 934.97
Synonym 1,2-Benzisothiazol-3(2H)-one, 1,1-dioxide, calcium salt, hydrate (4:2:7), 1,2-Benzisothiazol-3(2H)-one, 1,1-dioxide, calcium salt, hydrate (4:7) (9CI), Calcium 1,2-benzisothiazolin-3-one 1,1-dioxide hydrate, Calcium o-benzolsulfimide hydrate, Calcium saccharin hydrate
IUPAC navn dicalcium;1,1-dioxo-1,2-benzothiazol-2-id-3-one;heptahydrate
SMIL O.O.O.O.O.O.O.[Ca+2].[Ca+2].O=C1[N-]S(=O)(=O)c2ccccc12.O=C3[N-]S(=O)(=O)c4ccccc34.O=C5[N-]S(=O)(=O)c6ccccc56.O=C7[N-]S(=O)(=O)c8ccccc78
Tekniske data
Kemisk navn eller materiale Saccharin Calcium
Mængde 10 g
InChI formel InChI=1S/4C7H5NO3S.2Ca.7H2O/c4*9-7-5-3-1-2-4-6(5)12(10,11)8-7;;;;;;;;;/h4*1-4H,(H,8,9);;;7*1H2/q;;;;2*+2;;;;;;;/p-4
Formel vægt 933.9639