Få mere at vide
Saccharin-13C6, TRC
Artikelnummer. 17969744
Skift visning
Mængde:
0.5 mg
1 mg
2.5 mg
Pakningsstørrelse:
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17969744 1 mg -
17900105 0.5 mg 0.5mg
17967714 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17969744 Leverandør Toronto Research Chemicals Leverandørnr. TRCS0808021MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1286479-01-3
Molekylær formel C13C6H5NO3S
Molekylvægt (g/mol) 189.14
Synonym 1,2-(Benzisothiazol-13C6)-3(2H)-one 1,1-Dioxide, 1,2-Dihydro-2 -ketobenzisosulfonazole-13C6, Benzoic Sulfimide-13C6, o-Benzosulfimide-13C6, Benzosulfimide-13C6, Garantose-13C6, Glucid-13C6, Gluside-13C6, NSC 5349-13C6, Saccharimide-13C6, o-Sulfobenzimide-13C6;
IUPAC navn 1,1-dioxo-1,2-benzothiazol-3-one
SMIL O=C1NS([13C]2=[13CH][13CH]=[13CH][13CH]=[13C]21)(=O)=O
Tekniske data
Kemisk navn eller materiale Saccharin-13C6
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C7H5NO3S/c9-7-5-3-1-2-4-6(5)12(10,11)8-7/h1-4H,(H,8,9)/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vægt 189.0191
Procent renhed >95%