Få mere at vide
Sabinene, MedChemExpress
Beskrivelse
Sabinene is a perfume additive which is being explored as the component for the next generation aircraft fuel.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Sabinene |
| Fysisk form | Liquid |
| CAS | 3387-41-5 |
| Farve | Colorless |
| Mængde | 500 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C10H16 |
| Opløselighedsinformation | DMSO : 33.33 mg/mL (244.66 mM; Need ultrasonic) |
| SMIL | CC(C12CCC(C1C2)=C)C |
| Molekylvægt (g/mol) | 136.23 |
| Vis mere |