Få mere at vide
Sabinene Hydrate, TRC
Artikelnummer. 30424422
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424422 1 g
30428260 100 mg
30426981 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30424422 Leverandør Toronto Research Chemicals Leverandørnr. TRCS8075851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 546-79-2
Molekylær formel C10 H18 O
Molekylvægt (g/mol) 154.25
Synonym Bicyclo[3.1.0]hexan-2-ol, 2-methyl-5-(1-methylethyl)-, 4-Thujanol (6CI,7CI,8CI), 2-Methyl-5-(1-methylethyl)bicyclo[3.1.0]hexan-2-ol, 5-Isopropyl-2-methylbicyclo[3.1.0]hexan-2-ol, Sabinene hydrate, 4-Thujanol
IUPAC navn 4-methyl-1-propan-2-ylbicyclo[3.1.0]hexan-4-ol
SMIL CC(C)C12CCC(C)(O)C1C2
Tekniske data
Kemisk navn eller materiale 4-Thujanol
Mængde 1 g
InChI formel InChI=1S/C10H18O/c1-7(2)10-5-4-9(3,11)8(10)6-10/h7-8,11H,4-6H2,1-3H3
Formel vægt 154.1358