Learn More
SAAL1 Antibody - Azide and BSA Free, R&D Systems™
Product Code. 18615510 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18615510 0.02 mL 0.02mL
18679240 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18615510 Supplier Novus Biologicals Supplier No. NBP2936230.02ml
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody
SAAL1 Polyclonal antibody specifically detects SAAL1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen SAAL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias FLJ41463, protein SAAL1, serum amyloid A-like 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 175-474 of human SAAL1 (NP_612430.2). ERIQEHPAIYDSICFIMSSSTNVDLLVKVGEVVDKLFDLDEKLMLEWVRNGAAQPLDQPQEESEEQPVFRLVPCILEAAKQVRSENPEWLDVYMHILQLLTTVDDGIQAIVHCPDTGKDIWNLLFDLVCHEFCQSDDPPIILQEQKTVLASVFSVLSAIYASQTEQEYLKIEKVDLPLIDSLIRVLQNMEQCQKKPENSAESNTEETKRTDLTQDDFHLKILKDILCEFLSNIFQALTKETVAQGVKEGQLSKQKCSSAFQNLLPFYSPVVEDFIKILREVDKALADDLEKNFPSLKVQT
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Direct Reversal of DNA Damage
Primary or Secondary Primary
Gene ID (Entrez) 113174
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus