missing translation for 'onlineSavingsMsg'
Få mere at vide

Brady™ S3100 Sign and Label Printer with BWS Suite

Artikelnummer. 31842973
Skift visning
Klik for at se tilgængelige muligheder
Tastaturtype:
AZERTY
QWERTY
QWERTZ
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Tastaturtype
31842973 QWERTZ
31842974 QWERTY
31842970 AZERTY
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31842973 Leverandør Brady™ Leverandørnr. 198583

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

The easy-to-use Brady Printer S3100 sign and label printer is designed to meet a variety of your facility identification needs, helping you save time so you can get back to work.

  • Create the labels you need, when you need them without the wait with do-it-yourself label creation
  • Ideal for multi-user areas in your facility, anyone can simply walk up, create labels and print right on the spot
  • The built-in keyboard and touchscreen interface make label creation a breeze
  • Label creation is even faster with the quick setup and print speeds that come with print smart technology
  • Prints on a variety of material options so you can create signs and labels for a variety of applications
  • Ships with a license for the Brady Workstation suite of your preference: Product and Wire Identification, Safety and Facility Identification, or Lab Identification, enabling easy creation of a wide range of labels and signs.

Tekniske data

Produkttype Printer
Farve Blue, Gray
Display Type Color LCD Touchscreen
Til brug med (udstyr) B30 Series Thermal Transfer Tapes and Ribbons, B30 Series Thermal Transfer Tapes
Materiale Plastic
Print teknologi Thermal Transfer
Længde (engelsk) 10.25 in.
Bredde (engelsk) 12 in.
Længde (metrisk) 260.35 mm
Bredde (metrisk) 304.80 mm
Højde (engelsk) 9.5 in.
Højde (metrisk) 241.30 mm
Dimensioner (L x B x H) 260.35 x 304.80 x 241.30 mm
Autoklaverbar Not Autoclavable
Sterilitet Non-sterile
Farveevne Monocolour
Skærmstørrelse 155.44 x 91.95 mm
Inkluderer S3100 Printer, Power Cord, USB Cable, Cleaning Kit, Cutter Cleaning Tool, Quick Start Guide, Stylus, Mouse and a license for Brady Workstation Software suites
Sprog Bulgarian, Chinese (Simplified), Chinese (Traditional), Croatian, Czech, Danish, Dutch, English(US), Estonian, Finnish, French, German, Hungarian, Italian, Korean, Norwegian, Polish, Portuguese, Romanian, Russian, Slovak, Slovenian, Spanish
Hukommelse 256 MB
Model- S3100-QZ-BWS
Udskrivningshastighed 76.00 mm/sec.
Opløsning 300 dpi
Konnektortyper Ethernet, USB
Vægt (engelsk) 20 lb.
Vægt (metrisk) 9.07 kg
Certificeringer/compliance UL, cUL, CE, WEEE, RoHS, RCM (Australia/New Zealand), EAC (Russia, Belarus, and Kazakhstan), China: CCC-RoHS
Til brug med (applikation) Asset tracking, Cold temperature labelling, Component and equipment labelling, Electrical labelling, Exit and egress marking, Facility identification, Floor marking, General identification, Hazard communication identification, Lean and 5S labelling, Lockout tagout, Pipe and valve marking, Safety identification, Temperature indicating labelling
Mængde 1 Ea.
garanti 1 Year
Spænding 110/240 V
Tastaturtype QWERTZ
Alternativt varenummer S3100-QZ-BWS
Stiktype AC Adaptor
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.